ProKlenz® ONE High Performance Alkaline Cleaners
Supplier: Steris
1421D3
77531-596EA 23407.38 CAD
77531-596
ProKlenz® ONE High Performance Alkaline Cleaners
ProKlenz® ONE Cleaner is a proprietary blend of sodium hydroxide, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in clean-in-place (CIP) and clean-out-of-place (COP).
  • Productivity - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP) and clean-out-of-place (COP)
  • Easy to validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Efficacy - Delivers microbial control in critical manufacturing processes with disinfectant, virucidal and bioilm removal capabilities
  • Globally availability - Manufactured with biodegradable ingredients to support harmonization of cleaning processes
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
ProKlenz® ONE High Performance Alkaline Cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement and other industries requiring an effective validatable cleaner.
Biofilm Removal: ProKlenz® ONE Alkaline Cleaner is effective for biofilm removal when used at a 5% concentration of product per gallon of water on a pre-cleaned, non-porous, nonfood contact surface at 60 °C for a contact time of 10 minutes.
Disinfection: ProKlenz® ONE Alkaline Cleaner is effective against Staphylococcus aureus, Salmonella enterica, and Pseudomonas aeruginosa when used at a 1% concentration of product per gallon of water in 5% blood serum and 250 ppm of hard water at 60 °C for a contact time of 5 minutes.
Virucidal Activity: ProKlenz® ONE Alkaline Cleaner is effective against Poliovirus (type 1) when used at a 1% concentration of product per gallon of water in 5% blood serum and 250 ppm of hard water at 60°C for a contact time of 10 minutes.