Reusable Steel Pans
Supplier: Mettler Toledo
13954 214462
CA11278-148PK 161.31 CAD
CA11278-148 11278-060
Reusable Steel Pans
Trays/Dishes/Pots Laboratory Trays/Dishes
Reusuable steel pan can easily be washed and reused many times.
  • Robust
  • Solid steel sample pan
  • Dishwasher proof
It is environment-friendly and cost-effective.
For use with HC103, HE53, HE73, HS153 and HX204 model moisture analyzer.
Delivery information: Each packs contains 3 pans.