ASTM Class 4 Precision Weight Sets
Supplier: Mettler Toledo
11124066 11124068 11124070 11124072 11124074 11124080 11124084 11124067 11124069 11124071 11124073 11124075 11124081 11124085
CA97021-866EA 691.03 CAD
CA97021-866 CA97021-868 CA97021-870 CA97021-872 CA97021-874 CA97021-876 CA97021-878 CA97021-880 CA97021-882 CA97021-884 CA97021-886 CA97021-888 CA97021-890 CA97021-892
ASTM Class 4 Precision Weight Sets
Weights Calibration Weights
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
  • 316 stainless steel
  • Low magnetic permeability
  • High corrosion resistance
  • High gloss polish
  • ASTM Class 4
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.