Hanna Instruments pH Maintenance Solutions
Supplier: HANNA INSTRUMENTS CANADA CA
Hanna Instruments
HI54710-11 HI54710-11
CA470350-702PK 164.17 CAD
470350-702 CA470350-702
Hanna Instruments pH Maintenance Solutions
Environmental Test Kits
This is a complete set of all the solutions required for routine pH measurement.
  • 500 ml bottles
  • pH 4.01, 7.01 and 10.01 technical calibration solutions
  • Electrode storage solution and cleaning solution
The set includes pH 4.01, 7.01 and 10.01 technical calibration buffer that are NIST traceable and supplied with a certificate of analysis.
Ordering information: pH meter sold separately.