Revolve™ two-in-one mop covers, the first sustainable covers made for the cleanroom industry, are made from 100% upcycled polyester and are a part of Texwipe’s Revolve™ line of sustainable products.
- Integrated polyester cover/pad design allows replacing covers and pads in a one-step process
- Integrated design offers high sorption capacity; ideal for spill control, cleaning, and solution application
- Revolve™ processing provides low levels of ions, Non-Volatile Residues (NVRs), particles, and fibers
- For use in critical cleaning applications and environments
- ISO Class 2 to 7, Class 1 to 100,000, EU Grade A to D
- Gamma irradiated to a SAL of 10⁻⁶ according to AAMI guidelines
- Autoclave safe
Revolve™ mop covers contribute to a company's sustainability goals, reducing the amount of post-consumer water bottles going into landfills and reducing carbon emission levels.
Applications include mopping and cleaning large and small surfaces; applying and removing disinfectants; spill control; cleaning with solvents such as isopropyl alcohol (IPA), ethanol, acetone, and degreasers. Appropriate for use with temperatures less than 205 °C (400 °F).
Packaging: Covers are packed 10 per inner bag (triple bagged), 10 bags per case.