Human Recombinant SCYL1 (GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
S04-35G-100 S04-35G-1000
77563-424EA 1624.24 CAD
77563-424 77563-426
Human Recombinant SCYL1 (GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human SCYL1 (1-556) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice