Mouse Recombinant ROCK2 (His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
R11M-11H-10 R11M-11H-50 R11M-11H-500
77563-108EA 549.84 CAD
77563-108 77563-110 77563-112
Mouse Recombinant ROCK2 (His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant mouse ROCK2 (5-554) was expressed using baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice