Dog Recombinant RIPK1 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
R07D-11G-10 R07D-11G-50 R07D-11G-500
77562-972EA 549.84 CAD
77562-972 77562-974 77562-976
Dog Recombinant RIPK1 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant dog RIPK1 (1-327) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice