Human Recombinant KRAS (G12C) (from E. coli)
Supplier: Sino Biological
R06-32DU-100 R06-32DU-1000 R06-32DU-20
77562-882EA 913.24 CAD
77562-882 77562-884 77562-886
Human Recombinant KRAS (G12C) (from E. coli)
Proteins and Peptides
Recombinant human tag-free KRAS (G12C) (2-186) was expressed in E. coli cells. KRAS (G12C) is constitutively active.
  • Shipping temperature: Dry ice