Human Recombinant KRAS (G12C, His tag) (from E. coli)
Supplier: Sino Biological
R06-32DH-100 R06-32DH-1000 R06-32DH-20
77562-870EA 913.24 CAD
77562-870 77562-872 77562-874
Human Recombinant KRAS (G12C, His tag) (from E. coli)
Proteins and Peptides
Recombinant human KRAS (G12C) (2-186) was expressed in E.coli cells using an N-terminal His tag. The KRAS (G12C) mutant is constitutively active.
  • Shipping temperature: Dry ice