Human Recombinant RET (His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
R02-11H-10 R02-11H-50 R02-11H-500
77562-590EA 549.84 CAD
77562-590 77562-592 77562-594
Human Recombinant RET (His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human RET (658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice