Human Recombinant RET (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
R02-11G-500 R02-11G-50
77562-588EA 4168.04 CAD
77562-588 77562-586
Human Recombinant RET (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human RET (658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice