Human Recombinant PDE3A (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P91-31G-50 P91-31G-500
77562-418EA 2272.04 CAD
77562-418 77562-420
Human Recombinant PDE3A (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human PDE3A (669-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice