Human Recombinant AMPK (A1/B2/G3, His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P83-10G-500 P83-10G-10 P83-10G-50
77562-348EA 5921.84 CAD
77562-348 77562-344 77562-346
Human Recombinant AMPK (A1/B2/G3, His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human AMPK (combination of A1/B2/G3 subunits) was expressed by baculovirus in Sf9 insect cells using the N-terminal GST and C-terminal His tags.
  • Shipping temperature: Dry ice