Human Recombinant PKN1/PRK1 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P70-11G-10 P70-11G-50 P70-11G-500
77562-210EA 549.84 CAD
77562-210 77562-212 77562-214
Human Recombinant PKN1/PRK1 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human PKN1/PRK1 (289-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice