Human Recombinant AMPK (A1/B1/G3, His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P56-10H-50 P56-10H-500
77562-060EA 3267.44 CAD
77562-060 77562-062
Human Recombinant AMPK (A1/B1/G3, His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human AMPK (combination of A1/B1/G3 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tags.
  • Shipping temperature: Dry ice