Human Recombinant AMPK (A1/B2/G1, His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P50-10H-50 P50-10H-500
77561-944EA 3267.44 CAD
77561-944 77561-946
Human Recombinant AMPK (A1/B2/G1, His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human AMPK (combination of A1/B2/G1 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tags.
  • Shipping temperature: Dry ice