Human Recombinant PTPN12 (PTP-PEST, GST tag), Active (from E. coli)
Supplier: Sino Biological
P39-21G-50 P39-21G-500
77561-762EA 1308.24 CAD
77561-762 77561-764
Human Recombinant PTPN12 (PTP-PEST, GST tag), Active (from E. coli)
Proteins and Peptides
Recombinant human PTPN12 (1-355) was expressed in E. coli cells using a N-terminal GST tag.
  • Shipping temperature: Dry ice