Human Recombinant PIM2 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P36-10G-50 P36-10G-500
77561-676EA 1845.44 CAD
77561-676 77561-678
Human Recombinant PIM2 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human PIM2 was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
  • Shipping temperature: Dry ice