Human Recombinant PI3K (p65 alpha, His tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P31-31H-100
P31-31H-1000
77561-568EA
1292.44
CAD
77561-568
77561-570
Human Recombinant PI3K (p65 alpha, His tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human PI3K (p65 alpha) (1-571) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
- Shipping temperature: Dry ice