Human Recombinant PI3K (p85 beta, His tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P31-30BH-100 P31-30BH-1000
77561-550EA 1292.44 CAD
77561-550 77561-552
Human Recombinant PI3K (p85 beta, His tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human PI3K (p85β) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice