Human Recombinant PAD3 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P312-310CG-10 P312-310CG-50 P312-310CG-500
77561-522EA 534.04 CAD
77561-522 77561-524 77561-526
Human Recombinant PAD3 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human PAD3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice