Human Recombinant PI3K (p110 alpha (E542K, His tag)/p85 alpha, His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P27-122DH-50 P27-122DH-500
77561-416EA 1829.64 CAD
77561-416 77561-418
Human Recombinant PI3K (p110 alpha (E542K, His tag)/p85 alpha, His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human p110 alpha (E542K) mutant subunit and human p85 alpha wild-type subunit were co-expressed by baculovirus in Sf9 insect cells using an N-terminal His tag on both proteins.
  • Shipping temperature: Dry ice