Human Recombinant PPP2R1A (His tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P162-30H-100 P162-30H-1000 P162-30H-20
77561-244EA 1292.44 CAD
77561-244 77561-246 77561-248
Human Recombinant PPP2R1A (His tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human PPP2R1A was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice