Human Recombinant PPP2R1B (GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P162-30BG-100 P162-30BG-1000 P162-30BG-20
77561-232EA 1292.44 CAD
77561-232 77561-234 77561-236
Human Recombinant PPP2R1B (GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human PPP2R1B was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice