Human Recombinant PCNA (His tag) (from E. coli)
Supplier: Sino Biological
P11-30H-100 P11-30H-1000
77561-100EA 1624.24 CAD
77561-100 77561-102
Human Recombinant PCNA (His tag) (from E. coli)
Proteins and Peptides
Recombinant full length human PCNA was expressed by E. coli cells using an N-terminal His tag.
  • Shipping temperature: Dry ice