Human Recombinant SHIP2 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P09-20G-10 P09-20G-50 P09-20G-500
77561-058EA 597.24 CAD
77561-058 77561-060 77561-062
Human Recombinant SHIP2 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human SHIP2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice