Recombinant GST (from E. coli)
Supplier: Sino Biological
G52-30U-100 G52-30U-1000
77558-004EA 376.04 CAD
77558-004 77558-006
Recombinant GST (from E. coli)
Proteins and Peptides
Recombinant artificial sequences GST (1-226) was expressed in E. coli cells and purified on glutathione column.
  • Shipping temperature: Dry ice