Human Recombinant GRK6 (GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
G04-10G-50 G04-10G-500
77557-844EA 1845.44 CAD
77557-844 77557-846
Human Recombinant GRK6 (GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length human GRK6 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice