Human Recombinant FLT3 (Y591-V592insVDFREYEYD F691L, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
F12-12LG-10 F12-12LG-50 F12-12LG-500
77557-700EA 534.04 CAD
77557-700 77557-702 77557-704
Human Recombinant FLT3 (Y591-V592insVDFREYEYD F691L, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human FLT3 (Y591-V592insVDFREYEYD F691L) (571-993) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. This FLT3-ITD mutant has 9 amino acids “VDFREYEYD” repeated between the sites Y591 and V592 within the juxtamembrane domain.
  • Shipping temperature: Dry ice