Human Recombinant FGFR3 (GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
F06-11G-500 F06-11G-50
77557-528EA 4168.04 CAD
77557-528 77557-526
Human Recombinant FGFR3 (GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human FGFR3 (397-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice