Human Recombinant FGFR2 (C491F, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
F05-12IG-10 F05-12IG-50 F05-12IG-500
77557-464EA 534.04 CAD
77557-464 77557-466 77557-468
Human Recombinant FGFR2 (C491F, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human FGFR2 (C491F) (285-end) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice