Human Recombinant HER2 (V956R, His tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E27-12PH-10 E27-12PH-50 E27-12PH-500
77557-142EA 534.04 CAD
77557-142 77557-144 77557-146
Human Recombinant HER2 (V956R, His tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human HER2 (V956R) (677-1027) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice