Human Recombinant HER2 (R896C, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E27-122KG-10 E27-122KG-50 E27-122KG-500
77557-118EA 534.04 CAD
77557-118 77557-120 77557-122
Human Recombinant HER2 (R896C, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human HER2 (R896C) (676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice