Human Recombinant EPHB2 (GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E22-11G-50 E22-11G-500
77557-008EA 1845.44 CAD
77557-008 77557-010
Human Recombinant EPHB2 (GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human EPHB2 (570-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice