Human Recombinant EPHA8 (GST tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
E20-35G-100 E20-35G-1000
77556-988EA 1292.44 CAD
77556-988 77556-990
Human Recombinant EPHA8 (GST tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human EPHA8 (565-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice