Human Recombinant EPHA4 (Full length, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E16-11BG-10 E16-11BG-50 E16-11BG-500
77556-964EA 549.84 CAD
77556-964 77556-966 77556-968
Human Recombinant EPHA4 (Full length, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human EPHA4 (570-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice