Human Recombinant EGFR (T790M C797S L858R R999A, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E10-122R2G-10 E10-122R2G-50 E10-122R2G-500
77556-576EA 534.04 CAD
77556-576 77556-578 77556-580
Human Recombinant EGFR (T790M C797S L858R R999A, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human EGFR (T790M C797S L858R R999A) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice