Human Recombinant EGFR (K716A T790M C797S L858R, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E10-122P2G-10 E10-122P2G-50 E10-122P2G-500
77556-558EA 534.04 CAD
77556-558 77556-560 77556-562
Human Recombinant EGFR (K716A T790M C797S L858R, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human EGFR (K716A T790M C797S L858R) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice