Human Recombinant EGFR (d747-749, His tag), Active with mCherry (from Baculovirus (Insect cells))
Supplier: Sino Biological
E10-122MLH-10 E10-122MLH-50 E10-122MLH-500
77556-528EA 660.44 CAD
77556-528 77556-530 77556-532
Human Recombinant EGFR (d747-749, His tag), Active with mCherry (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human EGFR (d747-749) (668-end) with a C-terminal mCherry tag was expressed by baculovirus in Sf9 insect cells using an N-terminal His(10) tag.
  • Shipping temperature: Dry ice