Human Recombinant EGFR (d746-750, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E10-122JG-10 E10-122JG-50 E10-122JG-500
77556-480EA 534.04 CAD
77556-480 77556-482 77556-484
Human Recombinant EGFR (d746-750, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human EGFR (d746-750) (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice