Human Recombinant EGFR (His tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E10-112H-10 E10-112H-50 E10-112H-500
77556-336EA 549.84 CAD
77556-336 77556-338 77556-340
Human Recombinant EGFR (His tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human EGFR (668-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice