Human Recombinant DNMT3L (GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
D353-380CG-10 D353-380CG-50 D353-380CG-500
77556-250EA 660.44 CAD
77556-250 77556-252 77556-254
Human Recombinant DNMT3L (GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length human DNMT3L was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice