Human Recombinant DOT1L (KMT4, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
D344-381G-10 D344-381G-50 D344-381G-500
77556-232EA 660.44 CAD
77556-232 77556-234 77556-236
Human Recombinant DOT1L (KMT4, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human DOT1L (KMT4) (1-435) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice