Human Recombinant DGKQ (GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
D29-10G-10 D29-10G-50 D29-10G-500
77556-190EA 628.84 CAD
77556-190 77556-192 77556-194
Human Recombinant DGKQ (GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length human DGKQ was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice