Human Recombinant DGKI (His tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
D27-10BH-10 D27-10BH-50 D27-10BH-500
77556-160EA 628.84 CAD
77556-160 77556-162 77556-164
Human Recombinant DGKI (His tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length human DGKI was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice