Human Recombinant PDGFR Alpha (GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
P12-18G-50 P12-18G-500
77561-142EA 1845.44 CAD
77561-142 77561-144
Human Recombinant PDGFR Alpha (GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human PDGFRalpha (550 to end) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice