Human Recombinant PAK6 (GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
P06-10G-50 P06-10G-500
77560-982EA 1845.44 CAD
77560-982 77560-984
Human Recombinant PAK6 (GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length Human PAK6 was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice