Human Recombinant OAS1 (His Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
O711-30H-100 O711-30H-1000 O711-30H-20
77560-880EA 897.44 CAD
77560-880 77560-882 77560-884
Human Recombinant OAS1 (His Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length Human OAS1 was expressed by Baculovirus in Sf9 insect cells using an N to terminal His tag.
  • Shipping temperature: Dry ice