Human Recombinant NEK4 (GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
N06-10G-10 N06-10G-50 N06-10G-500
77560-514EA 549.84 CAD
77560-514 77560-516 77560-518
Human Recombinant NEK4 (GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length Human NEK4 was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice